Learn More
Wheaton™ Shaker Flasks With Four Baffles Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Wheaton™ 354237
 View more versions of this product
Catalog No. 10-042-8B
  Check Availability
Add to cart
Specifications
Description
  • For use with orbital or reciprocating shakers
  • Baffles improve aeration and mixing for microbial and bacterial cultures
  • Break resistant lip for use with stopper, cotton plug or film cover
Description & Specifications
Specifications
Yes
Style: Shaker
67mm
4.33 in.
Pinched
Borosilicate Glass
Baffled
2.63 in.
Orbital or reciprocating shakers
11cm
125mL