Wheaton™ Socorex Acura™ Multichannel Micropipetters Available on GSA/VA Contract for Federal Government customers only.
Fast and easy calibration reduces pipetter downtime
Manufacturer: Wheaton™ W810604
 View more versions of this product
Catalog No. 13-709-44
  Check Availability
Add to Cart
    Specifications
    Accuracy ±1%
    Compatible Tips Wheaton precision micropipette tip, universal fit tip
    Autoclavable Yes
    Number of Channels 8
    Volume (Metric) 40 to 350μL
    Increments 0.4μL
    Warranty 3 Years
    • Easy, swift-set calibration requires no additional tools beyond a balance and distilled water
    • Improved volume display can be seen easily during use, without changing your hand position
    • Tip ejector is positioned for use by either hand, even when wearing gloves
    • Autoclavable to 121°C (248°F), fully assembled
  • Description & Specifications
Related Products
Product Content Feedback