Wheaton™ Socorex Calibra™ 852 Digital Multichannel Micropipetter Available on GSA/VA Contract for Federal Government customers only.
Combines mechanical precision, comfort and ease of use
Manufacturer: Wheaton™ 8511882
 View more versions of this product
Catalog No. 13-702-4
  Check Availability
Add to Cart
    Specifications
    Autoclavable Yes
    Increments 1μL
    Material Stainless Steel
    Tip Style 200μL
    UV Monitoring Yes
    Warranty 3 Years
    Number of Channels 12
    Volume (Metric) 20 to 200μL
  • Description & Specifications
Related Products
Product Content Feedback