Wheaton™ Socorex Positive-Displacement Micropipetters
Provides positive displacement of sample in precision capillary tube
Manufacturer: Wheaton™ 851303
 View all options for this product
Catalog No. 13-707-43
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Number of Channels 1
    Product Type Socorex™ positive displacement micropipetter
    Volume (Metric) 10 to 50μL
    For Use With (Application) Interchangeable plunger and Glass capillary
    Warranty 3 Years
    • Step-wise volume adjustment
    • Interchangeable plunger and glass capillary
    • Matching color codes on pipette, plunger and capillary
    • Stainless-steel, PTFE-tipped plunger
    • Ideal for use with high-density, viscous, foaming or volatile liquids
  • Description & Specifications
Related Products
Product Content Feedback