Learn More
DWK Life Sciences Wheaton™ Square Media Bottles with Standard Caps
Click to view available options
Capacity (Metric):
250 mL
1000 mL
2000 mL
Description
- For storage of media, laboratory reagents and biological buffers
- Made of Eastman Tritan Eastar Copolyester PETG that is resin used for single use labware and medical devices
- With standard blue polypropylene caps
- Molded graduation markings for reference filling
- Gamma irradiated sterilized to SAL 10-6
- Cleanroom packaged for sterile production with multi-barrier elastomer packaging
- Resin meets ISO 10993 and/or USP Class VI biocompatibility requirements and is Food Contact Status compliant
Specifications
Capacity (Metric) | 250 mL |
Material | PETG |