Learn More
DWK Life Sciences Wheaton™ Stainless Steel Microtest Manifold
Make dispensing easier and quicker by selecting a Wheaton Stainless Steel manifold dispensing system.
Manufacturer:  DWK Life Sciences 851388
 View more versions of this product
Catalog No. 02-912-802
Add to cart
Description
Description
  • Unique dispensing system enables performance of sample inoculations and cell feedings faster and more efficiently under sterile conditions
  • Ideal for use with 24 and 96 well microplates
Specifications
Specifications
12
Microplates
Straight
Microtest Manifold
Yes
Stainless Steel