Learn More
DWK Life Sciences Stainless Steel Microtest Manifold
Make dispensing easier and quicker by selecting a Stainless Steel manifold dispensing system.
Supplier: DWK Life Sciences 851389
Description
- Unique dispensing system enables performance of sample inoculations and cell feedings faster and more efficiently under sterile conditions
- Ideal for use with 24 and 96 well microplates
Specifications
Microtest Manifold | |
Angled | |
Microplates |
Autoclavable | |
Stainless Steel | |
12 |