Learn More
Wheaton™ Sterile Vented Caps
Fit spinner flasks and roller bottles with 45mm finish
Manufacturer:  Wheaton™ W240752
 View more versions of this product
Catalog No. 22-288-527
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
No
Polypropylene
Screw
Glass spinner flasks or roller bottles with 45mm screw thread, allows gas exchange while preventing liquid, water or contaminants from passing through the filter
Cap
Green
45mm
Style: Vented
12
Sterile