Learn More
Wheaton™ Stoppers for Serum Bottles and Vials Available on GSA/VA Contract for Federal Government customers only.
Provide airtight covers
Manufacturer:  Wheaton™ W224100173
 View more versions of this product
Catalog No. 06-447J
  Check Availability
Add to cart
Specifications
Description
  • Five styles, in red natural rubber† or gray butyl
  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
Description & Specifications
Specifications
Yes
2cm
45 Durometer
Butyl
0.7 in.
Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials
Gray
Straight Plug Stopper