Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Snap-on Stopper
SDP

Provide airtight covers

Supplier:  DWK Life Sciences 224100180

 View more versions of this product

Catalog No. 06-447K


Edge
Add to Cart

Description

Description

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications

Specifications

20 mm
Yes
2 cm
Red
45 Durometer
Snap-On Stopper
13 mm
Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials
Rubber
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit