Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Sub Surface Grab Sampler II
SDP

For accessing fluid flow in deep manholes and hard-to-reach subsurface sampling areas

Supplier:  DWK Life Sciences 990400

 View more versions of this product

Catalog No. 15-437-2

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Includes: 1000mL narrow-mouth sample bottle with PTFE-lined cap

Description

Description

For accessing fluid flow in deep manholes and hard-to-reach subsurface sampling areas

  • Consists of an inert polypropylene head with stainless steel fittings and an aluminum pole assembly with golden alodine finish
  • Cable at top of handle lifts spring-loaded plunger from bottle opening and allows liquid to enter through four 15.9mm (0.625 in.) diameter hole

Specifications

Specifications

Subsurface Sampler
Polypropylene
1 L
EPA
3.66 m
1000mL narrow mouth sample bottle, cap
33.8 oz.
Golden alodine
12 ft.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit