Learn More
DWK Life Sciences Wheaton™ Tamper-Evident Caps
Contents cannot be accessed without removing cap or leaving evidence of puncture or insert
Manufacturer:  DWK Life Sciences W224199
 View more versions of this product
Catalog No. 03-084-130
  Check Availability
Add to cart
Description
Description
  • Use with Wheaton Uni-Dose Bottles and Vials
  • Three-piece Pulp / PVDC / PE lined aluminum caps have unique metal inserts
  • A metal tab with a rolled safety edge “perks up” to provide visual proof of tight seal
  • Caps remove quickly and easily in one piece
  • Cat. No. 03-084-130 is stamped inside “For Oral Use”
  • Attach using Wheaton Crimper
Specifications
Specifications
ISO 9001
Rolled safety edge
Uni dose bottle and vial
Pulp/PVDC/Polyethylene
5000 Cs.