Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Tenbroeck Tissue Grinder
With expanded reservoir and pouring lip
$297.00 - $407.00
Catalog Number | Mfr. No. | Capacity | Length (English) | Length (English) Tube | Tube Diameter (English) Outside | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Length (English) | Length (English) Tube | Tube Diameter (English) Outside | Price | Quantity & Availability | ||
08-414-18
|
DWK Life Sciences
357421 |
0.03 oz., 1mL | 5.51 in. | 1.8 in. | 0.43 in. |
Case of 2 for $297.00
|
|
||
08-414-13A
|
DWK Life Sciences
357422 |
0.06 oz., 2mL | 5.51 in. | 1.9 in. | 0.43 in. |
Case of 2 for $394.00
|
|
||
08-414-13B
|
DWK Life Sciences
357424 |
0.23 oz., 7mL | 7.4 in. | 3.2 in. | 0.62 in. |
Case of 2 for $407.00
|
|
||
08-414-13C
|
DWK Life Sciences
357426 |
0.5 oz., 15mL | 9.8 in. | 3.7 in. | 0.86 in. |
Case of 2 for $402.00
|
|
||
Description
With expanded reservoir and pouring lip
- Precision manufactured from Wheaton-33™ low extractable borosilicate glass
- Can be operated by hand or slow-speed motor
- Hollow handle permits packing with ice to reduce heat buildup
- Clearance between pestle and tube is approximately 0.12mm
Specifications
Borosilicate Glass | |
Tenbroeck Tissue Grinder |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title