Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Tenbroeck Tissue Grinder

With expanded reservoir and pouring lip

Manufacturer:  DWK Life Sciences 357426

 View more versions of this product

Catalog No. 08-414-13C


Add to cart

Description

Description

With expanded reservoir and pouring lip

  • Precision manufactured from Wheaton-33™ low extractable borosilicate glass
  • Can be operated by hand or slow-speed motor
  • Hollow handle permits packing with ice to reduce heat buildup
  • Clearance between pestle and tube is approximately 0.12mm

Specifications

Specifications

0.5 oz.
Borosilicate Glass
9.8 in.
3.7 in.
0.86 in.
0.5 oz., 15mL
15mL
Tenbroeck tissue grinder
250mm
94mm
22mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit