Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences™ Wheaton™ Thin Flange Stopper for Serum Bottles and Vials

Provide airtight covers for your serum bottles and vials with this thin flange stopper.
$259.00
Specifications
Description | Snap-on |
---|---|
Diameter (Metric) Inner | 2 cm |
Autoclavable | Autoclavable |
Color | Gray |
Material | Chlorobutyl Rubber |
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
01-549-775
![]() |
DWK Life Sciences™
W224100190 |
Snap-on |
Case of 300 for $259.00
|
|
|||||
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are ideal for freeze-dry applications
Specifications
Snap-on | |
Autoclavable | |
Chlorobutyl Rubber | |
Ribbed Cone | |
20 | |
Serum Bottles and Vials 1.3 cm I.D. and 2 cm O.D. |
2 cm | |
Gray | |
Thin Flange Stopper | |
Non-sterile | |
Lyophilization | |
300 Stoppers/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title