Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ TLC Developing Chambers

Ground glass covers securely fit chambers to reduce solvent evaporation

Manufacturer:  DWK Life Sciences 276845

 View more versions of this product

Catalog No. 06-411-53

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Glass cover

Description

Description

  • Holds plates up to 20 x 20cm in size
  • Made from soda-lime glass
  • Autoclavable
Includes glass cover

Specifications

Specifications

Yes
60 mm
10.5 cm
Borosilicate Glass
2.36 in.
4.1 in.
Glass cover
TLC Developing Chamber
Documents
Promotions

Promotions

  • DWK Life Sciences (Wheaton)Supplier Diversity Partner , DWK Life Sciences (Kimble)
    Buy $250 or more in DWK Life Sciences products, get a limited edition YOUtility Bottle at no additional cost.

    07-31-2019

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit