Learn More
DWK Life Sciences Wheaton™ TLC Developing Chambers
Ground glass covers securely fit chambers to reduce solvent evaporation
Specifications
Autoclavable Yes
View More Specs
Includes: Glass cover
Products 1
Catalog Number Mfr. No. Height (English) Height (Metric) Includes Material Price Quantity & Availability  
Catalog Number Mfr. No. Height (English) Height (Metric) Includes Material Price Quantity & Availability  
06-411-53 View Documents DWK Life Sciences
276845
4.1 in. 10.5 cm Glass cover Borosilicate Glass This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Holds plates up to 20 x 20cm in size
  • Made from soda-lime glass
  • Autoclavable
Includes glass cover
Specifications
Specifications
Yes