Wheaton™ TLC Sample Recovery Tubes
Manufacturer: Wheaton™ 277930
 View more versions of this product
Catalog No. 14-209-90
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Material Glass and PTFE
    Product Type Sample recovery tube
    Includes Case of Grease-free Wheaton connection
    For Use With (Application) Removal of sample from a developed TLC plate by vacuum
    Pore Size 40 to 60μ
    Thread Size 13 to 425 UNF
  • Description & Specifications
Related Products
Product Content Feedback