Wheaton™ Trypsinizing Flasks Available on GSA/VA Contract for Federal Government customers only.

Deep fluted sides enhance agitation

Manufacturer: Wheaton™ 355395

 View more versions of this product

Catalog No. 10-042-9D

  Check Availability
Add to Cart

    Specifications

    Graduated No
    Closure Size 38mm
    Closure Type Screw
    Capacity (Metric) 300mL
    For Use With (Application) Used for forming cell suspensions through Trypsin digestion of connective Proteins
    Style Erlenmeyer Trypsinizing
    Thread Size 430
    Material Borosilicate Glass
    Autoclavable Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit