Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
DWK Life Sciences Wheaton™ Unlined Aluminum Open-Top Seals
Open-top seal exposes stopper for sample retrieval with a syringe
Manufacturer:  DWK Life Sciences 22417607
 View more versions of this product
Catalog No. 11-126-20
Description
Description
  • Unlined
  • Closure Type: Seal
Specifications
Specifications
Open Top
Green
Case of 1000
Aluminum
Seal
11mm