Wheaton™ Unlined Aluminum Open-Top Seals Available on GSA/VA Contract for Federal Government customers only.
Open-top seal exposes stopper for sample retrieval with a syringe
Manufacturer: Wheaton™ 22417701
 View more versions of this product
Catalog No. 06-449-21
  Check Availability
Add to Cart
    Specifications
    Closure Type Open Top
    Thread Size 13mm
    Closure Material Aluminum
    Color Natural
    Quantity 1000 Case
    Type Seal
    Material Closure Aluminum
  • Description & Specifications
Related Products
Product Content Feedback