Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ V Vial™ with Open-Top Screw Cap

Conical interior bottoms are designed to provide total downward drainage and maximum retrieval of contents by syringe

Manufacturer:  DWK Life Sciences W986359NG

 View more versions of this product

Catalog No. 03-410-038


Add to cart

Includes: Silicone-lined, black phenolic screw caps faced with PTFE.

Description

Description

Made of “320” Type I glass for light-sensitive materials.

Specifications

Specifications

USP Type I
Open top screw cap
65mm
NextGen™ V Vial
Amber
Borosilicate Glass
PTFE/Silicone
Phenolic
Ideal for small scale reactions, centrifugation, storage, packaging and shipping
Screw
1.4 dr.
20mm
5mL
Conical V
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit