Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Wheaton™ V Vial™ with Solid-Top Screw Cap, Clear Available on GSA/VA Contract for Federal Government customers only.
Conical interior bottoms are designed to provide total downward drainage and maximum retrieval of contents by syringe
Manufacturer:  Wheaton™ W986274NG
 View more versions of this product
Catalog No. 22-021813
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
1.0mL
13-425
Clear
NextGen™ V Vial
Yes
Phenolic
PTFE/14B Rubber
13-425
0.3 dr.
Screw cap
1.0mL (0.03 oz.)
13mm
Low Extractable Borosilicate Glass
1mL
44mm
Ideal for small scale reactions, centrifugation, storage, packaging and shipping
Conical V
Screw