Learn More
DWK Life Sciences Wheaton™ Volumetric Modules for Acura™ Electro 926 Regular Shaft Length Pipets
Permanently store performance data in instrument's memory
Manufacturer:  DWK Life Sciences W870116
 View more versions of this product
Catalog No. 03-410-181
  Check Availability
Add to cart
Description
Description
  • Fit other volumetric assemblies offered separately
Specifications
Specifications
±1.2%
Ergonomic
0.05μL
200μL
0.5 to 10μL
200μL Pipette tip
Acura™ Electro™ 926XS regular shaft length single channel pipette
Regular Length
1
0.5 to 10μL