Learn More
DWK Life Sciences Volumetric Modules for Acura™ Electro 926 Regular Shaft Length Pipets
Permanently store performance data in instrument's memory
$90.64 - $124.63
Specifications
Finger Style Ergonomic
For Use With (Equipment) Acura™ Electro™ 926XS regular shaft length single channel pipette
Length (English) Regular Length
Number of Channels 1
View More Specs
Products 8
Catalog Number Mfr. No. Accuracy Volume (Metric) Increments Compatible Tips Price Quantity & Availability  
Catalog Number Mfr. No. Accuracy Volume (Metric) Increments Compatible Tips Price Quantity & Availability  
03-410-179
View Documents Promotion Details
DWK Life Sciences
W870112
±2.5% 0.1 to 2 μL 0.01μL Ultra 10μL pipette tip
Case for $112.27
Add to cart
03-410-180
View Documents Promotion Details
DWK Life Sciences
W870114
±1.2% 0.5 to 10 μL 0.05μL Ultra 10μL pipette tip
Case for $124.63
Add to cart
03-410-181
View Documents Promotion Details
DWK Life Sciences
W870116
±1.2% 0.5 to 10 μL 0.05μL 200μL Pipette tip
Case for $124.63
Add to cart
03-410-182
View Documents Promotion Details
DWK Life Sciences
W870118
±1.2% 1 to 20 μL 0.1μL 200μL Pipette tip
Case for $91.67
Add to cart
03-410-184
View Documents Promotion Details
DWK Life Sciences
W870122
±1% 5 to 100 μL 0.5μL 200μL Pipette tip
Case for $91.67
Add to cart
03-410-185
View Documents Promotion Details
DWK Life Sciences
W870124
±1% 10 to 200 μL 1μL 200μL Pipette tip
Case for $90.64
Add to cart
03-410-183
View Documents Promotion Details
DWK Life Sciences
W870120
±1% 2.5 to 50 μL 0.25μL 200μL Pipette tip
Case for $91.67
Add to cart
03-410-186
View Documents Promotion Details
DWK Life Sciences
W870126
±0.8% 50 to 1000 μL 5μL 1000μL Pipette tip
Case for $91.67
Add to cart
Description
Description
  • Fit other volumetric assemblies offered separately
Specifications
Specifications
Ergonomic
Acura™ Electro™ 926XS regular shaft length single channel pipette
Regular Length
1