Learn More
DWK Life Sciences Wheaton™ Volumetric Modules for Acura™ Electro 926 XS and 936 Single Channel Pipets
Interchangeable modules to fit different volumetric assemblies
Manufacturer:  DWK Life Sciences W870022
 View more versions of this product
Catalog No. 03-410-176
Add to cart
Description
Description
  • For reduced shaft length and single channel macrovolume pipets
  • One control unit fits a choice of 27 volumetric assemblies available separately
Specifications
Specifications
±1%
Ergonomic
0.5μL
200μL
5 to 100μL
200μL Pipette tip
Acura™ Electro™ 926XS reduced shaft length single channel pipette
Reduced Length
1
5 to 100μL