Learn More
DWK Life Sciences Wheaton™ Volumetric Modules for Acura™ Electro 926 XS and 936 Single Channel Pipets
Interchangeable modules to fit different volumetric assemblies
Manufacturer:  DWK Life Sciences W870024
 View more versions of this product
Catalog No. 03-410-177
  Check Availability
Add to cart
Description
Description
  • For reduced shaft length and single channel macrovolume pipets
  • One control unit fits a choice of 27 volumetric assemblies available separately
Specifications
Specifications
±1%
Ergonomic
1μL
200μL
10 to 200μL
200μL Pipette tip
Acura™ Electro™ 926XS reduced shaft length single channel pipette
Reduced Length
1
10 to 200μL