Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Foam/LDPE Liners

Click to view available options
Neck Finish:
15-415
20-410
24-400
24-410
28-400
28-410
33-400
33-430
Closure Liner:
Foam/LDPE
Polyethylene Foam

For general storage and packaging applications

  • Lined with foam polyethylene
  • Excellent barrier to aqueous solutions
  • Repeat sealability

Closure Liner Foam/LDPE
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Color White
Material Polypropylene
No. per Case 200 Cs.
Neck Finish 24-410