Learn More
Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners
Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
Manufacturer:  Wheaton™ 239279
 View more versions of this product
Catalog No. 02-984-20
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
ISO 9001
White
144 Cs.
22-400
Foam Polyethylene
Polypropylene
Glass bottle or vial