Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners Available on GSA/VA Contract for Federal Government customers only.

Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products

Manufacturer: Wheaton™ 239288

 View more versions of this product

Catalog No. 02-984-44

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ISO 9001
    Closure Liner Foam Polyethylene
    Color White
    For Use With Glass bottle or vial
    Material Polypropylene
    Neck Finish 48-400
    No. per Case 72 Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit