Wheaton™ White Polypropylene Caps with Polyvinyl Liners

Manufacturer: Wheaton™ 239212

 View all options for this product

Catalog No. 02-984-29


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Certifications/Compliance ISO 9001
    Closure Liner Poly/Vinyl
    Color White
    For Use With Round bottle
    Material Polypropylene
    Neck Finish 33-400
    No. per Case 144 Cs.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit