Learn More
Wheaton™ White Polypropylene Caps with Polyvinyl Liners Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Wheaton™ 239218
 View more versions of this product
Catalog No. 02-984-48
  Check Availability
Add to cart
Specifications
Description
  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier
Description & Specifications
Specifications
ISO 9001
White
72 Cs.
Round bottle
Poly/Vinyl
Polypropylene
Polypropylene Caps
58-400