Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners Available on GSA/VA Contract for Federal Government customers only.

Excellent for environmental samples, pharmaceuticals and diagnostic reagents

Manufacturer: Wheaton™ 239243

 View more versions of this product

Catalog No. 02-911-233

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ISO 9001
    Closure Liner PTFE/Polyethylene
    Color White
    For Use With Glass bottle or vial
    Material Polypropylene
    Neck Finish 63-400
    No. per Case 48 Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit