Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners Available on GSA/VA Contract for Federal Government customers only.
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
Manufacturer:  DWK Life Sciences 239233
 View more versions of this product
Catalog No. 02-984-22
  Check Availability
Add to cart
Description
Description
  • PTFE faced foamed polyethylene liner offers chemical resistance
  • Polyethylene foam provides compressibility and sealing properties
  • Good for high purity chemicals, strong acids, and solvents
  • For usewith glass bottles or vials
  • Polypropylene
  • Screw cap
  • White
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ISO 9001
White
144 Cs.
24-400
PTFE/Polyethylene
Polypropylene
Glass bottle or vial