Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
70-400
89-400
Material:
Polypropylene
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Description
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
- PTFE faced foamed polyethylene liner offers chemical resistance
- Polyethylene foam provides compressibility and sealing properties
- Good for high purity chemicals, strong acids, and solvents
- For usewith glass bottles or vials
- Polypropylene
- Screw cap
- White
Specifications
Closure Liner | PTFE/Polyethylene |
For Use With (Equipment) | Glass bottle or vial |
Color | White |
Material | Polypropylene |
No. per Case | 72 Cs. |
Neck Finish | 43-400 |
Certifications/Compliance | ISO 9001 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction