Learn More
DWK Life Sciences Wheaton™ White Polypropylene Open-Top Caps w/PTFE Faced Silicone Liners for E-Z Ex-Traction™ Vials Available on GSA/VA Contract for Federal Government customers only.
Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals
Manufacturer:  DWK Life Sciences W240846
 View more versions of this product
Catalog No. 02-927-121
Add to cart
Description
Description
Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ISO 9001
White
100 Cs.
Glass bottle or vial
24-400
PTFE/Silicone
Polypropylene
100/Cs.
Yes