Learn More
Amerex™ Wheeled Fire Extinguishers: Regular Dry Chemical
Wheeled, dry chemical fire extinguisher
Supplier:  Amerex™ 471
 View more versions of this product
Catalog No. 19-860-548
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Semi-pneumatic wheels
  • Coast Guard approved
  • Contains a siliconized sodium bicarbonate-based dry chemical with free flowing and non-caking additives
  • Economical Class B and C protection with lower initial cost and recharging
  • Chemical smothers fires in flammable liquids and pressurized gases and is electrically nonconductive
Specifications
Specifications
Steel-wheeled Fire Extinguisher
68 kg
1.9 cm
53 sec.
132 cm
50 ft.
15 m
30 to 40 ft.
-65° to +120°F
240B:C
207 kg
71 cm
150 lb.
0.75 in.
45 x 28 x 52 in. (114 x 71 x 132 cm)
52 in.
45 in.
114 cm
471
Regular dry chemical
-54°C to +49°C
457 lb.
28 in.