Learn More
Amerex™ Wheeled Fire Extinguishers: Regular Dry Chemical
Wheeled, dry chemical fire extinguisher
Supplier: Amerex™ 15504
Description
- Semi-pneumatic wheels
- Coast Guard approved
- Contains a siliconized sodium bicarbonate-based dry chemical with free flowing and non-caking additives
- Economical Class B and C protection with lower initial cost and recharging
- Chemical smothers fires in flammable liquids and pressurized gases and is electrically nonconductive
Specifications
Steel-wheeled Fire Extinguisher | |
159 kg | |
2.54 cm | |
70 sec. | |
145 cm | |
50 ft. | |
15 m | |
30 to 40 ft. | |
-65° to +120°F | |
320B:C | |
360 kg | |
88 cm |
350 lb. | |
1 in. | |
39 x 34 x 57 in. (99 x 88 x 145 cm) | |
57 in. | |
39 in. | |
99 cm | |
492 | |
Regular dry chemical | |
-54°C to +49°C | |
793 lb. | |
34 in. |