Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
While You Were Out Series
Never leave your substitute high and dry again.
$10.75
Specifications
Format | Book |
---|---|
For Use With (Application) | Concept crossword puzzles, formula word searches, challenging games within each discipline, answer sheets |
Min. Grade Level | 9 |
Catalog Number | Mfr. No. | Category | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Category | Product Type | Price | Quantity & Availability | ||||
S68108
![]() |
Science First![]() 2778 |
Physics | While You Were Out-Physical Science |
Each for $10.75
|
|
S68107
![]() |
Science First![]() 2774 |
Chemistry | While You Were Out-Chemistry |
Each for $10.75
|
|
Description
- For those emergency days when you cannot be in your classroom
- Provides science assignments that your substitute can do with your class
- Students can complete exercises by end of period
- Concept crossword puzzles
- Formula word searches
- Challenging games within each discipline
- Answer sheets
Specifications
Book | |
Concept crossword puzzles, formula word searches, challenging games within each discipline, answer sheets | |
9 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title