Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco Whirl-Pak™ Carrying Racks Available on GSA/VA Contract for Federal Government customers only.

For transporting and using sample-filled bags in laboratories

Manufacturer:  Nasco B00678WA

 View more versions of this product

Catalog No. 01-812-5D


  Check Availability
Add to cart

Description

Description

For transporting and using sample-filled bags in laboratories

  • Coated wire racks stand up to many lab materials
  • Double grid rack with second horizontal bar below top bar prevents bags from slipping sideways
  • 01-812-5G can be used for plate incubation by turning it on one side
  • Autoclavable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Yes
1 to 7 oz.
7.6cm
12.5 in.
Coated Wire
6.5 in.
Rack, Carrying
12.5 x 6.5 x 3 in. (31.8 x 16.5 x 7.6cm)
3 in.
Bags
31.8cm
15
16.5cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit