Learn More
Nasco Whirl-Pak™ Sampling Pole
Specially designed to hold any size bag, the holder uses a retainer ring to secure the bag firmly in place.
Manufacturer:  Nasco B01367WA
Catalog No. 01-812-92
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Even in a fast flowing stream, the bag will not come loose.
  • Adjustable fiberglass pole extends from 6 to 12 ft. (1.82 to 3.66 m), has a swinging end, and comes with three sizes of retainer rings to hold the bag.
  • Not autoclavable. Can be sanitized with a bleach and water mixture.
Specifications
Specifications
Polyethylene
Retainer ring
1.8 to 3.6 m
Sampling Pole
6 to 12 ft.