Learn More
3M™ White Disposable Visitor Lab Coats, Series 4400
Use for hygiene applications. The 3M Disposable Visitor Lab Coats series is made from lightweight polypropylene.
$294.66 - $335.43
Specifications
Closure Type Zipper
Color White
Cuff Style Knit
Material Polypropylene
Disposable Yes
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-624-128
View Documents
3M™
7000089710
Large
Case of 50 for $294.66
Add to cart
17-624-131
View Documents
3M™
7000089713
3X-Large
Case of 50 for $335.43
Add to cart
Description
Description
  • Breathable, lightweight material
  • Designed for light duty applications
  • Offers a barrier against non-hazardous particles and liquid splashes
  • Zip fastener
  • Anti-static treated
  • Knitted cuffs
  • 3M Catalog# 4440-3XL , 3M Catalog# 4440-L
Specifications
Specifications
Zipper
Knit
Yes
Serged
White
Polypropylene
Used in catering, clean rooms, electronics manufacturing, general maintenance, hygiene, laboratories