Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner
Commonly used for food, beverage, medical and chemical applications packed at less than 120° F. Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner are known for good impact strength and resistant to mild acids, alkalis, solvents, alcohols, oils and aqueous products.
 View more versions of this product
Catalog No. 02-912-291A
  Check Availability
Add to Cart
Alert
Not recommended for active hydrocarbons or bleaches.
Includes: Pulp/Vinyl liner
    Specifications
    Product Type Polyvinyl Lined Cap
    Closure Color White
    Closure Material Polypropylene
    Closure Type Screw
    Color White
    Material Polypropylene
    Liner Material Pulp/Vinyl
    Neck Finish 48-400
    Packaging Type Packed in 48 bags of 12
    Quantity 576 Each/Cs.
    Shape Round
    Polypropylene caps are:
    • Known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
    • Autoclavable
    Pulp/Vinyl liners are:
    • Made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
    • Commonly used for food, beverage, medical and chemical applications packed at less than 120° F., including mild acids, alkalis, solvents, alcohols, oils and aqueous products
  • Description & Specifications
Related Products
Product Content Feedback