Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

White Swan™ Essentials™ Women's Princess-Style Labcoat

Manufacturer:  White Swan META 47401W40

 View more versions of this product

Catalog No. 19-154-109


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Button
Cotton Poplin,Polyester
40
Female
46.5 in.
38.5 in.
118cm
Set-in
100.3cm
White
3
Stain release
Mock belt, single breasted, 5-button front
49 in.
124.46cm
97.79cm
Princess
39.5 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit