Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

White Swan™-Meta™ Maxima™ Fluid-Resistant Lab Coat with Knit Cuffs

Fluid resistant, with MetaGuard™ protection

Manufacturer:  White Swan META 11653 11 L

 View more versions of this product

Catalog No. 18-999-945D


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

OSHA
White, Slight Grayish
Polyester
Large
Adjustable self adhesive
42 to 44 in.
106.68 to 111.72cm
101.6cm
Polyester, Carbon Fiber
Adjustable self adhesive
Knit
3
No
Fluid
36 to 38 in.
91.44 to 96.52cm
40 in.
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit