Learn More
Brady™ White Tyvek™ Tags
Identify facility equipment to your exact specifications. Brady™ White Tyvek™ Tags are durable one-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 102035
 View more versions of this product
Catalog No. 19-112-958
Add to cart
Description
Description
  • Tyvek™ tags are a lightweight, tear proof and weather resistant alternative to cardstock tags
  • They provide more durability but still offer the flexibility to write your own custom message on the spot
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
Specifications
Specifications
White
Not laminated
Rectangle
2.125 in. x 4.25 in.
1000/Pack
Tyvek™