Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ White Tyvek™ Tags

Identify facility equipment to your exact specifications. Brady™ White Tyvek™ Tags are durable one-sided tags with a write-on area that accepts most marking pens.

$266.00 - $393.00

Specifications

Color White
Material Tyvek™
Label Type Not laminated
Shape Rectangle
View More Specs

Products 5
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-112-957 View Documents Brady™
102032
1.375 in. x 2.75 in. Pack of 1000 for $266.00
Add to cart
 
19-112-958 View Documents Brady™
102035
2.125 in. x 4.25 in. Pack of 1000 for $307.00
Add to cart
 
19-112-960 View Documents Brady™
102037
2.625 in. x 5.25 in. Pack of 1000 for $349.50
Add to cart
 
19-112-959 View Documents Brady™
102036
2.375 in. x 4.75 in. Pack of 1000 for $314.00
Add to cart
 
19-112-961 View Documents Brady™
102038
2.875 in. x 5.75 in. Pack of 1000 for $393.00
Add to cart
 
Description

Description

  • Tyvek™ tags are a lightweight, tear proof and weather resistant alternative to cardstock tags
  • They provide more durability but still offer the flexibility to write your own custom message on the spot
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
Specifications

Specifications

White
Tyvek™
Not laminated
Rectangle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit