Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Whitney™ Medical Solutions Broken Glass Keeper™ Collection and Disposal Container

Safely dispose of non-contaminated broken glass in sturdy, corrugated, clearly marked disposal container

Manufacturer:  Whitney™ Medical Solutions BH2008

 View more versions of this product

Catalog No. 14-375-263


Add to cart

Description

Description

Available in floor and bench-top models. Both models made in USA.

Floor Model

  • Sturdy corrugated container
  • Thick, clear plastic liner
  • Unique Reclose-a/Remove-a-Lid™ with extra wide opening
  • Inner flap on lid seals the lid securely for disposal
  • Ships and stores flat
  • Dimensions: 12W x 12D x 27 in.H (30.5 x 30.5 x 68.6cm)
  • Capacity: 16.6 gal. (63L)
Bench-Top Model
  • Made of sturdy paperboard with an internal plastic lining and webbed corners to contain moderate amounts of fluids
  • Self-locking tops keep contents safely enclosed
  • Bench-Top Keepers fold out to 6L x 6W x 10 in.H (15 x 15 x 25cm) and accommodate up to 5000cc of material
  • Autoclavable

Specifications

Specifications

63 L
Non-contaminated broken glass
Floor
16.6 gal.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit