Learn More
Whitney™ Medical Solutions eShield™ Sterile Digital Camera Cover
Used to cover digital cameras in sterile field. Whitney Medical Solutions™ eShield™ Smart Phone Cover enables the safe use of digital camera for photography in cleanroom and controlled environment.
Manufacturer:  Whitney™ Medical Solutions EC2200
Catalog No. 22-375-403
Add to cart
Includes: Set of adhesive rings for lens bezel
Description
Description
  • FDA approved sterile cover for digital cameras
  • Protects the sterile environment from contaminants on handheld devices
  • Constructed of ultra-clear polyethylene and allows for easy operation of the device controls with gloved hands
  • Enclosed adhesive lens rings adhere cover tightly across lens bezel for clear photos
  • Facilitates easy manipulation of camera controls through cover
  • Meets ASTM F1671 Viral Penetration Guidelines
  • Easily accommodates aseptic technique
  • Camera Brands: Apple Canon, Kodak, Nikon, Olympus, Panasonic, Sony and most other cameras
Specifications
Specifications
PE (Polyethylene) film
9 x 14 in.