Learn More
Allcells LLCSupplier Diversity Partner Whole Blood, Fresh, Syringe NaCitrate, 60mL DFS Item
Manufacturer:  Allcells LLCSupplier Diversity Partner WB, FR, 48ML (60ML), SNC
AllCells Whole blood (WB) is anti-coagulated using heparin, ACD:A, EDTA or sodium citrate per customer specification. Our standard anticoagulant is heparin (5 U/ml of WB). The volume of whole blood removed per donation may vary, but will not exceed 420 mL at any one time. Whole blood collection quantities are in volume increments of 60 mL up to a maximum of 420 mL however custom collections in increments of 10 ml are available upon request. Please contact us if smaller volumes are needed. WB can be collected in vacutainer tubes, syringes, or transfer packs.
Catalog No. 50-146-8742